Learn More
MSC SPI Threaded Ring Gage, M8 x 1.00, Metric, Class 6G, Go
Catalog No. 5000537529
Encompass
This threaded ring gage is designed for checking M8 x 1.00 metric threads. It features a Class 6G classification and is a 'Go' gage, ensuring precise measurement.
  • Class 6G
  • Oil hardened non-shrinking steel (OHNS)
  • NPL traceability certification included
Catalog No. 50-005-37529 Supplier MSC Supplier No. 75890921
This threaded ring gage is designed for checking M8 x 1.00 metric threads. It features a Class 6G classification and is a 'Go' gage, ensuring precise measurement.
  • Class 6G
  • Oil hardened non-shrinking steel (OHNS)
  • NPL traceability certification included