Learn More
MSC SPI V-Block, 13/16 in Capacity, 90° V Angle, Hardened Steel
Catalog No. 5000531512
Encompass
This V-Block set features a 13/16 in maximum capacity and a 90° V angle, making it suitable for precision workholding in various applications. Constructed from hardened, precision-ground tool steel, it ensures durability and accuracy. The set includes two blocks and two clamps, designed for reliable and repeatable fixturing.
  • Supplied as a 2-block set
  • Includes two clamps for secure fastening
  • Made from hardened and precision ground tool steel
  • Squareness and parallelism within 0.0003 in
  • Flatness within 0.0002 in
  • Numbered and matched pair to guarantee accuracy
  • Non-magnetic design
  • Hardness of 50-60 HRC
Catalog No. 50-005-31512 Supplier MSC Supplier No. 85687531
This V-Block set features a 13/16 in maximum capacity and a 90° V angle, making it suitable for precision workholding in various applications. Constructed from hardened, precision-ground tool steel, it ensures durability and accuracy. The set includes two blocks and two clamps, designed for reliable and repeatable fixturing.
  • Supplied as a 2-block set
  • Includes two clamps for secure fastening
  • Made from hardened and precision ground tool steel
  • Squareness and parallelism within 0.0003 in
  • Flatness within 0.0002 in
  • Numbered and matched pair to guarantee accuracy
  • Non-magnetic design
  • Hardness of 50-60 HRC