Learn More
MSC SPI V-Block, 3-1/16" Max Capacity, 90° V Angle, Matched Pair
Supplier:  MSC 02662880
Encompass
This V-Block provides stable and precise support for cylindrical workpieces during various industrial applications such as machining, inspection, and layout tasks. Designed with a 3-1/16 inch maximum capacity and a 90 degree V angle, it is constructed from durable cast iron to ensure high accuracy and long-term reliability. This product is supplied as a matched pair, ensuring consistent performance for critical operations.
  • 6 inch overall length
  • 2-7/16 inch overall width
  • 3-1/2 inch overall height
  • 3-1/16 inch maximum V capacity
  • 90 degree V angle
  • Constructed from cast iron
  • Provided as a matched pair
  • V parallelism of 0.0002 inch
  • V accuracy of 0.0002 inch
  • Non-magnetic design
Catalog No. 50-005-29813