Learn More
MSC SPI V-Block, Magnetic, 1-3/4 in Max Capacity, 90° V Angle
Catalog No. 5000514154
Encompass
This V-block is a precision tool made from steel, designed for holding cylindrical workpieces. It features a magnetic base for secure placement and offers accurate measurements for various applications.
  • Magnetic for secure placement
  • Made from durable steel
  • 1-3/4 in maximum capacity
  • 90° V angle
  • V parallelism of 10.00 micro m
  • Squareness per 1 inch of 10.00 micro m
  • Sold as an individual unit
Catalog No. 50-005-14154 Supplier MSC Supplier No. 07176761
This V-block is a precision tool made from steel, designed for holding cylindrical workpieces. It features a magnetic base for secure placement and offers accurate measurements for various applications.
  • Magnetic for secure placement
  • Made from durable steel
  • 1-3/4 in maximum capacity
  • 90° V angle
  • V parallelism of 10.00 micro m
  • Squareness per 1 inch of 10.00 micro m
  • Sold as an individual unit