Learn More
MSC SPI Vernier Caliper, 0 to 6 in, 0.001 in Accuracy, Stainless Steel
Catalog No. 5000528567
Encompass
This vernier caliper offers precise measurements from 0 to 6 inches (0 to 150 mm) with an accuracy of 0.001 inch (0.02 mm). Constructed from stainless steel, it provides durability and reliability for various applications. It features both inch and metric display units and includes fine adjustment capabilities for enhanced control during measurements.
  • Fine adjustment for precise control.
  • Includes a protective case for storage and transport.
  • Constructed from stainless steel for durability.
Catalog No. 50-005-28567 Supplier MSC Supplier No. 50318807
This vernier caliper offers precise measurements from 0 to 6 inches (0 to 150 mm) with an accuracy of 0.001 inch (0.02 mm). Constructed from stainless steel, it provides durability and reliability for various applications. It features both inch and metric display units and includes fine adjustment capabilities for enhanced control during measurements.
  • Fine adjustment for precise control.
  • Includes a protective case for storage and transport.
  • Constructed from stainless steel for durability.