Learn More
MSC SPI Vertical Dial Test Indicator, 0.008 in. Range, 0.0001 in. Graduation, 1 ea
Catalog No. 5000511665
Encompass
The SPI Vertical Dial Test Indicator is a high-precision instrument designed for accurate measurement in various machining and inspection applications. Featuring a fully jeweled movement, this indicator provides reliable, sensitive, and repeatable performance with universal positioning capabilities to suit diverse setup requirements.
  • Range of 0.008 in. with 0.0001 in. graduation
  • Easy to read 1-1/4 in. white dial with 0-4-0 reading
  • Contact point with 0.080 in. diameter and +/-100 degrees of pivot
  • Includes 5/32 in. and 3/8 in. diameter dovetail stems
  • Features a durable satin chrome finish
  • Supplied in a fitted case with a short form certificate of calibration
Catalog No. 50-005-11665 Supplier MSC Supplier No. 37543667
The SPI Vertical Dial Test Indicator is a high-precision instrument designed for accurate measurement in various machining and inspection applications. Featuring a fully jeweled movement, this indicator provides reliable, sensitive, and repeatable performance with universal positioning capabilities to suit diverse setup requirements.
  • Range of 0.008 in. with 0.0001 in. graduation
  • Easy to read 1-1/4 in. white dial with 0-4-0 reading
  • Contact point with 0.080 in. diameter and +/-100 degrees of pivot
  • Includes 5/32 in. and 3/8 in. diameter dovetail stems
  • Features a durable satin chrome finish
  • Supplied in a fitted case with a short form certificate of calibration