Learn More
MSC SPI Wire and Sheet Metal Gage, Steel, 3.25 in. Diameter, 1 ea.
Catalog No. 5000512453
Encompass
This wire and sheet metal gage is a precision measuring tool designed for checking material thickness. Constructed from durable steel, it provides reliable performance in industrial metalworking and inspection environments.
  • Manufactured from steel for enhanced durability
  • Designed for measuring thickness of wire and sheet metal
  • Suitable for use with non-ferrous metals
  • Measures 3.25 in. in diameter with 0.13 in. thickness
Catalog No. 50-005-12453 Supplier MSC Supplier No. 42614933
This wire and sheet metal gage is a precision measuring tool designed for checking material thickness. Constructed from durable steel, it provides reliable performance in industrial metalworking and inspection environments.
  • Manufactured from steel for enhanced durability
  • Designed for measuring thickness of wire and sheet metal
  • Suitable for use with non-ferrous metals
  • Measures 3.25 in. in diameter with 0.13 in. thickness