Learn More
Aviva Systems Biology Corp SPI1 Antibody - middle region (ARP38241_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38241_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SPI1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SPI1. Alternate names: OF, PU.1, SFPI1, SPI-1, SPI-AApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rat, Zebrafish
Catalog No. 50-014-56577