Learn More
Aviva Systems Biology Corp SPIB Antibody - C-terminal region (ARP31422_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP31422_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SPIB. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human SPIB. Alternate names: SPI-BApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-52827