Learn More
Aviva Systems Biology Corp SPIC Antibody - N-terminal region (ARP37892_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37892_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against SPIon Channel. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of mouse SPIC. Alternate names: Prf, Spi-C, C76795, AU019198ApplicationWB. Species reactivity: Mouse
Catalog No. 50-014-53096