Learn More
Aviva Systems Biology Corp SPIC Antibody - N-terminal region (ARP39881_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39881_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SPIC. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SPIC. Alternate names: MGC40611, SPI-CApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Rabbit, Rat
Catalog No. 50-014-57216