Learn More
Miles Scientific Spice Sample Preparation Cartridges 50/pack
SDP
Catalog No. NC0661618
Encompass_Preferred
UNIBOND(TM) C18 SPICE(TM) SAMPLE PREPARATION CARTRIDGES PKG 48. 3.10 LBS. Analtech SPICE Cartridges are prepacked with adsorbent and ready for use. Molded of medical grade polypropylene, each cartridge contains 1 cc of packing. A porous frit and outlet filter combination contain the packing and will remove sample particulate down to 10 microns. In normal use SPICE sample preparation cartridges are compatible with organic solvents and aqueous solutions. Precision Luer fittings provide leak proof liquid connections.
Catalog No. NC0661618 Supplier Miles Scientific Supplier No. S0110
UNIBOND(TM) C18 SPICE(TM) SAMPLE PREPARATION CARTRIDGES PKG 48. 3.10 LBS. Analtech SPICE Cartridges are prepacked with adsorbent and ready for use. Molded of medical grade polypropylene, each cartridge contains 1 cc of packing. A porous frit and outlet filter combination contain the packing and will remove sample particulate down to 10 microns. In normal use SPICE sample preparation cartridges are compatible with organic solvents and aqueous solutions. Precision Luer fittings provide leak proof liquid connections.