Learn More
Arrow Engineering SSS-55 Spider Support Stand
Catalog No. NC2088760
Encompass_Preferred
The Arrow Mixing Products SSS-55 commercial mixer stand fits vessels up to a 23″ inscribed circle,fits a 55 gallon drum(please note, the stand does NOT come with a bucket). Overall height of the Spider Support Stand is 26″ with a 5/8″ dia center rod gives you the ability to mount your mixer or stirrer directly to your solutions container.
This item is not returnable. View return policy
Catalog No. NC2088760 Supplier Arrow Engineering Supplier No. SSS55
The Arrow Mixing Products SSS-55 commercial mixer stand fits vessels up to a 23″ inscribed circle,fits a 55 gallon drum(please note, the stand does NOT come with a bucket). Overall height of the Spider Support Stand is 26″ with a 5/8″ dia center rod gives you the ability to mount your mixer or stirrer directly to your solutions container.