Learn More
Mitegen Spider for Vitrobot Dewar By Nanosoft
SDP
Catalog No. 5000495012
Encompass
The Nanosoft Spider for Vitrobot Dewar is a vital tool for cryo-EM sample preparation. Its design mimics the spider from the Thermo Fisher Vitrobot dewar set, ensuring seamless compatibility and equal performance. The spider is engineered to work with the styrofoam dewar set from the Vitrobot, making it a reliable addition to your cryo-vitrification workflow.
Catalog No. 50-004-95012 Supplier Mitegen Supplier No. MCEMNSSPDR01
The Nanosoft Spider for Vitrobot Dewar is a vital tool for cryo-EM sample preparation. Its design mimics the spider from the Thermo Fisher Vitrobot dewar set, ensuring seamless compatibility and equal performance. The spider is engineered to work with the styrofoam dewar set from the Vitrobot, making it a reliable addition to your cryo-vitrification workflow.