Learn More
Bellco Glass, Inc. Spigot, Up/Down Lever, 2/PK
SDP
Supplier:  Bellco Glass, Inc. 452800413
Encompass_Preferred
EZgrip carboy spigots have large levers that control two flow rates, one for continuous, locked flow, and the other for faster, manual flow. The spigot spout features 5/8" and 3/4" hose barbs, and a standard internal Luer tape to fit a male barb adapter. The spigot body has internal threads with a hard stop to prevent over-tightening which may damage the seal. Spigots come in a pack of 2. Spigots are made from polypropylene, which may be autoclaved at 15 PSI and 121° (250°) for 20 minutes.Two flow rates (continuous or manual)Made from Class VI autoclavable polypropyleneInternal threads with a hard stop to prevent over-tightening*Only compatible with EZgrip and EZBio Carboys. Thread: 1 1/8Material: PolypropyleneGasket Material: SiliconO-ring Material: SiliconLength: 1.5" (38 mm)Width: 2.9" (74 mm)Height: 4.0" (102 mm)
Catalog No. 50-255-0349