Learn More
Cayman Chemical Spike Glycoprotein (1147-1162) (SARS-CoV-2, SARS-CoV) (trifluoroacetate salt), Purity: ≥95%, 25 mg
SDP
Catalog No. 502917356
Encompass_Preferred
A SARS-CoV-2 spike glycoprotein-derived peptide; antibodies against this peptide neutralize SARS-CoV-2 and SARS-CoV pseudovirus particles in cellular assays
Catalog No. 50-291-7356 Supplier Cayman Chemical Supplier No. 3749125 MG
A SARS-CoV-2 spike glycoprotein-derived peptide; antibodies against this peptide neutralize SARS-CoV-2 and SARS-CoV pseudovirus particles in cellular assays