Learn More
Cayman Chemical Spike Glycoprotein 1147-1162
SDP
Catalog No. 502915639
Encompass_Preferred
A SARS-CoV-2 spike glycoprotein-derived peptide; antibodies against this peptide neutralize SARS-CoV-2 and SARS-CoV pseudovirus particles in cellular assays
Catalog No. 50-291-5639 Supplier Cayman Chemical Supplier No. 3749150 MG
A SARS-CoV-2 spike glycoprotein-derived peptide; antibodies against this peptide neutralize SARS-CoV-2 and SARS-CoV pseudovirus particles in cellular assays