Learn More
Cayman Chemical Spike Glycoprotein (1147-1162) (SARS-CoV-2, SARS-CoV) (trifluoroacetate salt), Purity: ≥95%, 5 mg
SDP
Catalog No. 502916137
Encompass_Preferred
A SARS-CoV-2 spike glycoprotein-derived peptide; antibodies against this peptide neutralize SARS-CoV-2 and SARS-CoV pseudovirus particles in cellular assays
Catalog No. 50-291-6137 Supplier Cayman Chemical Supplier No. 374915 MG
A SARS-CoV-2 spike glycoprotein-derived peptide; antibodies against this peptide neutralize SARS-CoV-2 and SARS-CoV pseudovirus particles in cellular assays