Learn More
Cayman Chemical SpIke InhIbItr Screeng Rea 1ea
SDP
Catalog No. 502945612
Encompass_Preferred
Spike Inhibitor Screening Reagent has been tested and formulated to work exclusively with Cayman’s SARS-CoV-2 Spike-ACE2 Interaction Inhibitor Screening Assay Kit
Catalog No. 50-294-5612 Supplier Cayman Chemical Supplier No. 4020561 EA
Spike Inhibitor Screening Reagent has been tested and formulated to work exclusively with Cayman’s SARS-CoV-2 Spike-ACE2 Interaction Inhibitor Screening Assay Kit