Learn More
ACROBiosystems Biotinylated SARS-CoV-2 Spike Trimer, His,Avitag (B.1.1.529/Omicron) (SPN-C82Ee) is the ectodomain of SARS-CoV-2 spike protein which contains AA Val 16 - Pro 1213 (Accession # QHD43416.1 ).
Supplier:  ACROBiosystems SPNC82EE25UG
Encompass_Preferred
It's been reported that SARS-CoV-2 can infect the human respiratory epithelial cells through interaction with the human ACE2 receptor. The spike protein is a large type I transmembrane protein containing two subunits, S1 and S2. S1 mainly contains a receptor binding domain (RBD), which is responsible for recognizing the cell surface receptor. S2 contains basic elements needed for the membrane fusion.The S protein plays key parts in the induction of neutralizing-antibody and T-cell responses, as well as protective immunity.
Catalog No. 50-223-1118