Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACROBiosystems Human Anti-SARS-CoV-2 Spike RBD Antibody-Coupled Magnetic Beads, 10 mg

Catalog No. 502370258
Encompass_Preferred

Pre-coupled magnetic beads coated with anti-SARS-CoV-2 Spike RBD antibody for specific immunocapture and biopanning applications. Supplied as a stabilized powder (10 mg, 5 mg x 2) formulated in PBS with 10% trehalose; store at -20 °C. Reconstitute to 1 mg/mL (5 mL for 10 mg) and avoid more than three freeze-thaw cycles. Non-sterile; ready for use after reconstitution.

  • Pre-coupled for specific capture of Spike RBD.
  • Uniform 2 μm particles for efficient capture and reproducible results.
  • High binding capacity: >192 pmol (10 μg) Spike RBD per mg beads.
  • Supplied as powder; reconstitute to 1 mg/mL (5 mL for 10 mg).
  • Formulated in PBS, pH 7.4, with 10% trehalose for stability.
  • Store at -20 °C and avoid more than three freeze-thaw cycles.
  • Non-sterile; intended for immunocapture and biopanning workflows.

Catalog No. 50-237-0258 Supplier ACROBiosystems Supplier No. MBSK01410MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Pre-coupled magnetic beads coated with anti-SARS-CoV-2 Spike RBD antibody for specific immunocapture and biopanning applications. Supplied as a stabilized powder (10 mg, 5 mg x 2) formulated in PBS with 10% trehalose; store at -20 °C. Reconstitute to 1 mg/mL (5 mL for 10 mg) and avoid more than three freeze-thaw cycles. Non-sterile; ready for use after reconstitution.

  • Pre-coupled for specific capture of Spike RBD.
  • Uniform 2 μm particles for efficient capture and reproducible results.
  • High binding capacity: >192 pmol (10 μg) Spike RBD per mg beads.
  • Supplied as powder; reconstitute to 1 mg/mL (5 mL for 10 mg).
  • Formulated in PBS, pH 7.4, with 10% trehalose for stability.
  • Store at -20 °C and avoid more than three freeze-thaw cycles.
  • Non-sterile; intended for immunocapture and biopanning workflows.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.