Learn More
ACROBiosystems ACROBiosystems SARS-CoV-2 Spike Trimer (T19R, V70F, FR157-158Del, A222V, W258L, K417N, L452R, T478K, D614G, P681R, D950N), His Tag
Catalog No. NC2010478
Encompass_Preferred
Recombinant Protein;500UG;SARS-CoV-2 Spike Trimer, His Tag (SPN-C52Ht) is expressed from human 293 cells (HEK293). It contains AA Val 16 - Pro 1213 (Accession # QHD43416.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 137.8 kDa.
Catalog No. NC2010478 Supplier ACROBiosystems Supplier No. SPNC52HT500UG
Recombinant Protein;500UG;SARS-CoV-2 Spike Trimer, His Tag (SPN-C52Ht) is expressed from human 293 cells (HEK293). It contains AA Val 16 - Pro 1213 (Accession # QHD43416.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 137.8 kDa.