Learn More
ACROBiosystems ACROBiosystems SARS-CoV-2 Spike RBD (L452Q, F490S), His Tag
Catalog No. NC2010462
Encompass_Preferred
Recombinant Protein;100UG;SARS-CoV-2 Spike RBD, His Tag (SPD-C5227) is expressed from human 293 cells (HEK293). It contains AA Arg 319 - Lys 537 (Accession # QHD43416.1 (L452Q, F490S)).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 26.5 kDa.
Catalog No. NC2010462 Supplier ACROBiosystems Supplier No. SPDC5227100UG
Recombinant Protein;100UG;SARS-CoV-2 Spike RBD, His Tag (SPD-C5227) is expressed from human 293 cells (HEK293). It contains AA Arg 319 - Lys 537 (Accession # QHD43416.1 (L452Q, F490S)).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 26.5 kDa.