Learn More
ACROBiosystems ACROBiosystems Biotinylated SARS-CoV-2 Spike RBD (L452Q, F490S), His,Avitag™
Supplier:  ACROBiosystems SPDC82EH200UG
Encompass_Preferred
Recombinant Protein;200UG;Biotinylated SARS-CoV-2 Spike RBD, His,Avitag (SPD-C82Eh) is expressed from human 293 cells (HEK293). It contains AA Arg 319 - Lys 537 (Accession # QHD43416.1 (L452Q, F490S)).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag (Avitag™). The protein has a calculated MW of 28.2 kDa.
Catalog No. NC2010459