Learn More
Cygnus Technologies Cygnus Technologies Spiking MVP, Viral Clearance Surrogate, BSL-1 Compatible, Non-Infectious, 1.5 mL
Catalog No. NC2109311
Encompass
The Cygnus Technologies Spiking MVP is a non-infectious viral clearance surrogate specifically designed for advanced viral clearance prediction in biopharmaceutical applications. Offering an economical alternative to live virus studies, this product mimics the physicochemical properties of the Minute Virus of Mice (MVM), facilitating robust process validation and enhancing biotherapeutic safety. Ideal for in-house spiking studies, it supports scientists in optimizing viral clearance across the biopharmaceutical lifecycle.
This item is not returnable. View return policy
Catalog No. NC2109311 Supplier Cygnus Technologies Supplier No. M2211
The Cygnus Technologies Spiking MVP is a non-infectious viral clearance surrogate specifically designed for advanced viral clearance prediction in biopharmaceutical applications. Offering an economical alternative to live virus studies, this product mimics the physicochemical properties of the Minute Virus of Mice (MVM), facilitating robust process validation and enhancing biotherapeutic safety. Ideal for in-house spiking studies, it supports scientists in optimizing viral clearance across the biopharmaceutical lifecycle.