Learn More
FyterTech Nonwovens Spilfyter™ Battery Acid Spill Kits
Catalog No. 19812523 Shop All NPS Corporation Products
Change view
Click to view available options
Battery Acid Spill Kit
Catalog No.
19-812-523 Battery Acid Spill Kit
1 options
Catalog No. 19-812-523 Supplier FyterTech Nonwovens Supplier No. 440133
Add to Cart
Add to Cart
Includes
KolorSafe™ Liquid Acid Neutralizer (19-021-888), 2 qt. (1.89L); one pair nitrile gloves; polyester apron; scoop with detachable scraper; ten 12 x 12 in. Hazmat Pads (19-121-018); Aqualockit™ Polymer, 1 oz. (28g); splash goggles; trigger sprayer; and disposal bag with twist tie
Ideal for quick response to leaking or spilled battery acid
Kits should be located in garages, forklift storage areas, batteries storage areas; also used for cleanup of dry cell batteries.
Specifications
Type Battery Acid Spill Kit
For Use With (Application) Quick response to leaking or spilled battery acid
Includes 2 quarts KOLOR-SAFE™ liquid acid neutralizer, 12 x 12 in. hazmat pads (10), 1 oz. AQUALOCKIT™ polymer, 1 pair nitrile gloves, 1 pair splash goggles, 1 poly apron, 1 trigger sprayer, 1 scoop with detachable scraper
Weight (English) 9 lb.
Weight (Metric) 4 kg