Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FyterTech Nonwovens Spilfyter™ Specialty Spill Control, Decontamination #4

Catalog No. 19066542 Shop All NPS Corporation Products
Change view
Click to view available options
Capacity (English):
55 gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English)
19-066-542 55 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-066-542 Supplier FyterTech Nonwovens Supplier No. 680044

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Use for base and caustic contaminants such as organic bases, alkali and caustic wastes.

Designed for on-site decontamination of personal protective equipment, tools and surfaces at hazardous material incidences or waste sites.
  • “Ready-to-Use” solutions requiring no dilution
  • Phosphate-free
  • 5 gallon Carboy
  • 47 lb.

Specifications

Capacity (English) 55 gal.
Capacity (Metric) 208 L
Type Decontamination Spill Kit
For Use With (Application) Base and caustic contaminants such as organic bases, alkali and caustic wastes
Includes 1 Decon solutions 1, 2, 3, 4 and 5 (in 5 gal. Carboys), 1 Poly drum, 55 gal., 1 poly sheeting, 10 x 30 ft., 6 hazmat docks, 1 containment pond, 1 pair of Nitrile gloves, 1 disposable bag and twist tie, 2 Poly sprayers, 3 gal. capacity, 3 decon wash brushes, 1 aqualockit™ shaker bottle, 32 oz., 1 roll ph paper, 1 pair of splash goggles
Weight (English) 47 lb.
Weight (Metric) 21 kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.