Learn More
FyterTech Nonwovens Spilfyter™ Disinfecting Wipe Kit Pro Refill
Catalog No. 19066556 Shop All NPS Corporation Products
Change view
Click to view available options
Packaging Quantity:
2 oz. Concentrate (6 in each refill)
Catalog No. Packaging Quantity
19-066-556 2 oz. Concentrate (6 in each refill)
1 options
This item is not returnable. View return policy
Catalog No. 19-066-556 Supplier FyterTech Nonwovens Supplier No. 74491EF
This item is not returnable. View return policy
Refill this easy-to-use solution featuring a professional disinfectant cleaner on the EPA List N for emerging pathogens. The Disinfecting Wipe Kit Pro helps you clean shared work spaces and equipment.
  • Easily refill your Disinfecting Wipe Kit Pro.
  • This simple to use kit delivers a professional solution featuring a convenient, pre-portioned disinfecting cleaner.
  • One-step cleaning and disinfecting solution is EPA-approved for Emerging Pathogens (List N EPA Reg No. 6836-365-5449).
  • Tested and confirmed 30-day efficacy once mixed and added to the wipes.
  • Each coreless wiper roll features 300 dry, Airlaid cellulose-based wipers (6 ¾ x 6 in.) that pull from the center – perfect for feeding wipers through the bucket lid.
  • Can be used to clean and disinfect all hard surfaces, equipment, tools and more.
  • Wipers easily absorb the disinfecting solution while remaining durable to handle your daily cleaning needs.
  • Refills include six wiper rolls and six disinfectant cleaner concentrates.
Specifications
Packaging Quantity 2 oz. Concentrate (6 in each refill)
Saturation Solution Quat Based (128 E-FECTicide from Multi Clean)
Sterility Non-sterile
pH Range 7 to 8