Learn More
FyterTech Nonwovens Spilfyter™ Grab and Go Neutralizer Spill Kits
Catalog No. 19066470 Shop All NPS Corporation Products
Change view
Click to view available options
Neutralizer Spill Kit
Catalog No.
19-066-470 Neutralizer Spill Kit
1 options
Do not use KolorSafe™ Liquid Base Neutralizer on any solution containing (in any form) metallic nitrates, cyanides, sulfides, strong oxidizers or hypochlorite (sodium or calcium) solutions since dangerous gases will be generated
Catalog No. 19-066-470 Supplier FyterTech Nonwovens Supplier No. 270002
Add to Cart
Add to Cart
Do not use KolorSafe™ Liquid Base Neutralizer on any solution containing (in any form) metallic nitrates, cyanides, sulfides, strong oxidizers or hypochlorite (sodium or calcium) solutions since dangerous gases will be generated
Includes
  • Liquid Acid Neutralizer 1 qt.
  • Liquid Base Neutralizer 1 qt.
  • 2# Aualockit™ Shaker bottle
  • Nitrile Gloves 1 pr.
  • Splash Goggles 1 pr.
  • Wisk-on Pan
  • Trigger Sprayer
  • Disposal Bag w/Twist Tie
Handle small acid or base spills quickly
  • Compact spill kit with everything needed for small acid/base spills
  • KolorSafe™ liquid neutralizers have color indicator for easy identification of neutralization
  • Plastic case with handle makes the kit easy to store and carry
Specifications
Type Neutralizer Spill Kit
Includes 1 qt. liquid acid neutralizer , 1 qt. liquid base neutralizer, 2 Aualockit™ shaker bottle, 1 pair nitrile gloves, 1 pair splash goggles wisk on pan trigger sprayer disposal bag with twist tie
Weight (English) 11 lb.
Weight (Metric) 5 kg