Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FyterTech Nonwovens Spilfyter™ Kollect-A-Kem™ Chemical Spill Control Center

Catalog No. 18999567 Shop All NPS Corporation Products
Change view
Click to view available options
Capacity (English):
8 gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English)
18-999-567 8 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 18-999-567 Supplier FyterTech Nonwovens Supplier No. 200800

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Space saving, plastic wall-mount spill center

Specifications

Capacity (English) 8 gal.
Capacity (Metric) 30.3 L
Type Chemical Spill Station
Includes (10) 8.5 x 17 in. pillows, (10) 12 x 12 in. pads, 2 pair nitrile gloves, 2 aprons, 2 goggles, 10 bags with ties, 1 plastic cabinet with clear hinged door
Weight (English) 18 lb.
Weight (Metric) 8 kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.