Learn More
FyterTech Nonwovens Spilfyter™ MERCSORB™ Sponges
Catalog No. 19812530 Shop All NPS Corporation Products
Change view
Click to view available options
Packaging Quantity:
12/Cs.
Catalog No. Packaging Quantity
19-812-530 12/Cs.
1 options
Catalog No. 19-812-530 Supplier FyterTech Nonwovens Supplier No. 520261
Sponges for cleaning up mercury spills
  • Replacement part for MERCSORB™ Amalgamation spill kit
  • Sponges work to absorb liquid from contaminated areas
Specifications
Type Mercury Sponge
For Use With (Application) Mercury spill cleanup, used as a replacement for Mercsorb amalgamation spill kit
Packaging Quantity 12/Cs.