Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Hazardous Disposal Bag

Catalog No. 19103126 Shop All Brady Corporation Products
Change view
Click to view available options
Capacity (English):
17 to 20 gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English)
19-103-126 17 to 20 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-103-126 Supplier Brady™ Supplier No. BAG104
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Replenish any spill kit with this hazardous materials disposal bag. Can also be purchased individually for custom kits.

  • Hazardous materials disposal bag with a 13-17 gallon capacity
  • Works well in just about any spill kit
  • This part is typically found in kits, but can be purchased individually for custom kits or as a replacement

Order Info

Minimum order quantity is 250.

Specifications

Certifications/Compliance ROHS Compatibility: Pass 2011/65/EU(Pass 2015/863)
Color Yellow
Capacity (English) 17 to 20 gal.
Material Plastic
Type Spill Kit Bag
For Use With (Application) Replacement bag for spill kits
Packaging Quantity 1 Ea.
Dimensions (L x W) 31 in. x 26 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.