Learn More
Marketlab Spill Clean Up Kit
Description
- 1 Pair of Vinyl Gloves
- Red Z Solidifier (10g Pouch)
- Scoop/Scraper
- Red Biohazard Waste Bag (Dimensions 24 in. x 24 in.)
- Twist Tie
- p.a.w.s Antimicrobial Hand Wipe (Dimensions: 5 in. x 8 in.)
Specifications
Specifications
| Type | Spill Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.