Description
Construction
- (10) Paper towels
- (2) Chloride antiseptic towelettes
- (1) Disinfectant cloth
- (1) Scoop
- (1) Spatula
- (1) Pair of nitrile exam gloves
- (1) Absorbent powder pack
- (1) Biohazard bag
- (1) Unmarked bag
- (2) Twist ties
Performance
- Protects user against direct contact with pathogens and infectious substances
- Assortment of cleaning tools and disposable personal protective equipment
Specifications
Specifications
| Material | Plastic |
| For Use With (Application) | Biohazard Spill Cleanup |