Learn More
Thomas Scientific Spill Containment Tray, 34 x 18 x 2"
SDP
Catalog No. NC1488779
Encompass
Protect the bottom of your laboratory cabinets with these containment trays. These trays are commonly manufactured with a 2" containment lip in case of spills. Laboratory spill trays are a minimum 3/16” thick, one piece stress free, virgin polyethylene with smooth interior walls and coved corners for easy cleaning. Trays conform to ASTM #1412 for corrosion resistance, and provide secondary containment complying with OSHA 1910.1450 and EPA 264.175 for chemical stockrooms/storerooms All trays are manufactured in the USA
Catalog No. NC1488779 Supplier Thomas Scientific Supplier No. 1185F78
Protect the bottom of your laboratory cabinets with these containment trays. These trays are commonly manufactured with a 2" containment lip in case of spills. Laboratory spill trays are a minimum 3/16” thick, one piece stress free, virgin polyethylene with smooth interior walls and coved corners for easy cleaning. Trays conform to ASTM #1412 for corrosion resistance, and provide secondary containment complying with OSHA 1910.1450 and EPA 264.175 for chemical stockrooms/storerooms All trays are manufactured in the USA