Learn More
California Pacific Lab & Consulting Secondary Spill Containment Tray, Polyethylene, 3/16" Thick, 3 x 3", 2" High
SDP
Catalog No. NC3785957
Encompass_Preferred
Secondary spill trays for safety, compliance and additional protection in your chemical storage areas. Catch drips, prevent spills and corrosion, and keep certain chemicals segregated by using a tray sysem. Polyethylene spill trays provide long lasting containment without the risk of corrosion or breakage. One-piece rotationally molded design offers drip compliance and protects cabinetry. -100% polyethylene construction. -Acid resistant / highly chemical resistant resin. -One-piece design. -Thick 3/16" walls are made of an impact-resistant plastic that resists breakage. -Interior corners are coved for easy cleaning. -Dishwasher safe. -Natural white color. -Made in the USA
Catalog No. NC3785957 Supplier California Pacific Lab & Consulting Supplier No. SCIPT520303201
Secondary spill trays for safety, compliance and additional protection in your chemical storage areas. Catch drips, prevent spills and corrosion, and keep certain chemicals segregated by using a tray sysem. Polyethylene spill trays provide long lasting containment without the risk of corrosion or breakage. One-piece rotationally molded design offers drip compliance and protects cabinetry. -100% polyethylene construction. -Acid resistant / highly chemical resistant resin. -One-piece design. -Thick 3/16" walls are made of an impact-resistant plastic that resists breakage. -Interior corners are coved for easy cleaning. -Dishwasher safe. -Natural white color. -Made in the USA