Learn More
Electron Microscopy Sciences Spill Control Tray
SDP
Catalog No. 5028839
Encompass_Preferred
The tray is made from vacuum formed polystyrene 1/2"(12.7cm) deep with an external flange.The overall dimensions are 23"x27" (58x69cm). The tray accommodates our Benchtop mat (Cat. #64446). These trays are very useful in the biology or chemistry labs.
Catalog No. 50-288-39 Supplier Electron Microscopy Sciences Supplier No. 64448
The tray is made from vacuum formed polystyrene 1/2"(12.7cm) deep with an external flange.The overall dimensions are 23"x27" (58x69cm). The tray accommodates our Benchtop mat (Cat. #64446). These trays are very useful in the biology or chemistry labs.