Learn More
CEM Corporation SPILL CUP CAVITY INSERT
SDP
Catalog No. NC9880656
Encompass
Standard polypropylene spill cup supplied with the Discover reactor. The spill cup inserts directly into cavity to protect the cavity and infrared sensor from damage due to spillage or ruptured reaction vials.
Catalog No. NC9880656 Supplier CEM Corporation Supplier No. 162425
Standard polypropylene spill cup supplied with the Discover reactor. The spill cup inserts directly into cavity to protect the cavity and infrared sensor from damage due to spillage or ruptured reaction vials.