Learn More
MSC Brady SPC Sorbents Hazardous Waste Bags, 17 Gal, Yellow
Catalog No. 502839774
Encompass
These hazardous waste bags from Brady SPC Sorbents are designed for safely containing and disposing of various hazardous materials. Each bag offers a 17 US gallon capacity and is constructed from durable 3 mil thick material, ensuring reliable containment for laboratory and industrial applications.
  • Pack of 250 bags for bulk supply
  • 17 US gallon capacity for ample waste containment
  • Yellow color for easy identification of hazardous waste
  • 3 mil thickness for enhanced durability and tear resistance
  • Dimensions of 26 in wide by 31 in high
  • Flat pack dispensing for convenient storage and access
Catalog No. 50-283-9774 Supplier MSC Supplier No. 78251204
These hazardous waste bags from Brady SPC Sorbents are designed for safely containing and disposing of various hazardous materials. Each bag offers a 17 US gallon capacity and is constructed from durable 3 mil thick material, ensuring reliable containment for laboratory and industrial applications.
  • Pack of 250 bags for bulk supply
  • 17 US gallon capacity for ample waste containment
  • Yellow color for easy identification of hazardous waste
  • 3 mil thickness for enhanced durability and tear resistance
  • Dimensions of 26 in wide by 31 in high
  • Flat pack dispensing for convenient storage and access