Learn More
Newcomer Supply Spill Response Kits, Polyform-F, 2 x 32 oz.
SDP
Catalog No. NC0779326
Encompass
Spill kit for formaldehyde and glutaraldehyde. Kit includes: Polyform-F, Safety goggles, gloves, waste bags & broom/pan.
Catalog No. NC0779326 Supplier Newcomer Supply Supplier No. ABSRKPF3202
Spill kit for formaldehyde and glutaraldehyde. Kit includes: Polyform-F, Safety goggles, gloves, waste bags & broom/pan.