Learn More
Chemglass Life Sciences Spill Tray, Polyethylene, 14"L x 14"W x 2"H
SDP
Catalog No. NC1154567
Encompass_Preferred
Spill Tray, Polyethylene,Constructed of chemical-resistant 3/16 thick virgin polyethylene. Trays are designed to contain any accidental spills.14"L x 14"W x 2"H
Catalog No. NC1154567 Supplier Chemglass Life Sciences Supplier No. CG1988A02
Spill Tray, Polyethylene,Constructed of chemical-resistant 3/16 thick virgin polyethylene. Trays are designed to contain any accidental spills.14"L x 14"W x 2"H