Learn More
Chemglass Life Sciences Spill Tray, Polyethylene, 19"L x 16"W x 2"H
SDP
Catalog No. 501531370
Encompass_Preferred
Spill Tray, Polyethylene, 19"L x 16"W x 2"H. Constructed of chemical-resistant 3/16 thick virgin polyethylene. Trays are designed to safely contain accidental spills. Unless otherwise noted, they are rotationally molded, one-piece design with smooth, coved corners; no welded joints that can leak, break off or trap contaminates.
Catalog No. 50-153-1370 Supplier Chemglass Life Sciences Supplier No. CG1988A06
Spill Tray, Polyethylene, 19"L x 16"W x 2"H. Constructed of chemical-resistant 3/16 thick virgin polyethylene. Trays are designed to safely contain accidental spills. Unless otherwise noted, they are rotationally molded, one-piece design with smooth, coved corners; no welded joints that can leak, break off or trap contaminates.