Learn More
Chemglass Life Sciences Spill Tray, Polyethylene, 38"L x 14"W x 2"H (Fits Most Flammable Cabinets)
SDP
Catalog No. 501531372
Encompass_Preferred
Spill Tray, Polyethylene, 38"L x 14"W x 2"H (Fits Most Flammable Cabinets). Constructed of chemical-resistant 3/16 thick virgin polyethylene except for CG-1988-02 and CG-1988-03, which are welded polypropylene. Trays are designed to safely contain accidental spills. Unless otherwise noted, they are rotationally molded, one-piece design with smooth, coved corners; no welded joints that can leak, break off or trap contaminates.
Catalog No. 50-153-1372 Supplier Chemglass Life Sciences Supplier No. CG1988A20
Spill Tray, Polyethylene, 38"L x 14"W x 2"H (Fits Most Flammable Cabinets). Constructed of chemical-resistant 3/16 thick virgin polyethylene except for CG-1988-02 and CG-1988-03, which are welded polypropylene. Trays are designed to safely contain accidental spills. Unless otherwise noted, they are rotationally molded, one-piece design with smooth, coved corners; no welded joints that can leak, break off or trap contaminates.