Learn More
Chemglass Life Sciences Spill Tray, Polyethylene, 31"L x 16"W x 2"H
SDP
Catalog No. NC1271759
Encompass_Preferred
Spill Tray, Polyethylene,Constructed of chemical-resistant 3/16 thick virgin polyethylene. Trays are designed to contain any accidental spills.31"L x 16"W x 2"H
Catalog No. NC1271759 Supplier Chemglass Life Sciences Supplier No. CG1988A16
Spill Tray, Polyethylene,Constructed of chemical-resistant 3/16 thick virgin polyethylene. Trays are designed to contain any accidental spills.31"L x 16"W x 2"H