Learn More
Chemglass Life Sciences Spill Tray, Polyethylene, 22"L x 19"W x 2"H (Fits 24" Cabinets)
SDP
Catalog No. NC1207427
Encompass_Preferred
Spill Tray, Polyethylene,Constructed of chemical-resistant 3/16 thick virgin polyethylene. Trays are designed to contain any accidental spills.22"L x 19"W x 2"H (Fits 24" Cabinets)
Catalog No. NC1207427 Supplier Chemglass Life Sciences Supplier No. CG1988A12
Spill Tray, Polyethylene,Constructed of chemical-resistant 3/16 thick virgin polyethylene. Trays are designed to contain any accidental spills.22"L x 19"W x 2"H (Fits 24" Cabinets)