Learn More
Chemglass Life Sciences Spill Tray, Polyethylene, Fits Reactor Frame: CG-1950-100, 17"L x 22"W x 2"H, Capacity: 6L
SDP
Catalog No. 50872455
Encompass_Preferred
Spill Tray, Polyethylene, Fits Reactor Frame: CG-1950-100, 17"L x 22"W x 2"H, Capacity: 6L. Acid-Resistant Safety Spill TraysEasy-to-Clean Coved Corners (unless otherwise noted)Great for Trays, Drawer and Shelf LinersCustom Sizes AvailableConstructed of chemical-resistant 3/16¿ thick virgin polyethylene. Trays are designed to safely contain accidental spills. Unless otherwise noted, they are rotationally molded, one-piece design with smooth, coved corners; no welded joints that can leak, break off or trap contaminates.
Catalog No. 50-872-455 Supplier Chemglass Life Sciences Supplier No. CG198801
Spill Tray, Polyethylene, Fits Reactor Frame: CG-1950-100, 17"L x 22"W x 2"H, Capacity: 6L. Acid-Resistant Safety Spill TraysEasy-to-Clean Coved Corners (unless otherwise noted)Great for Trays, Drawer and Shelf LinersCustom Sizes AvailableConstructed of chemical-resistant 3/16¿ thick virgin polyethylene. Trays are designed to safely contain accidental spills. Unless otherwise noted, they are rotationally molded, one-piece design with smooth, coved corners; no welded joints that can leak, break off or trap contaminates.