Learn More
Eagle SpillNEST™ Containment Pools
GREENER_CHOICE
Catalog No. 19999016 Shop All Eagle Manufacturing Products
Change view
Click to view available options
Material:
PVC Fabric
Catalog No. Material
19-999-016 PVC Fabric
1 options
Catalog No. 19-999-016 Supplier Eagle Supplier No. T8611
Add to Cart
Add to Cart
Drip Pads designed for outdoor leaks and spills and compatible with all fuels and hydrocarbons.
  • Each unit folds easily for storage
  • Refill pads are available
Specifications
Material PVC Fabric
Type Spill Containment Pallet
Weight Capacity Lightweight
Weight (English) 4 lb.
Weight (Metric) 1.8 kg