Learn More
Eagle SpillNEST™ Drip Pads
GREENER_CHOICE
Catalog No. 19999015 Shop All Eagle Manufacturing Products
Change view
Click to view available options
Material:
PVC Fabric
Catalog No. Material
19-999-015 PVC Fabric
1 options
Catalog No. 19-999-015 Supplier Eagle Supplier No. T8341
Add to Cart
Add to Cart
Designed to be used as durable, portable drip pan for outdoor leaks and spills and are compatible with all fuels and hydrocarbons.
  • Each unit folds easily for storage
  • Refill pads are available
Specifications
Material PVC Fabric
Type Spill Containment Pallet
Weight Capacity Medium-weight
Weight (English) 113 lb.
Weight (Metric) 51.26 kg