Learn More
Justrite™ Spillslope Steel Dispensing Shelf for all 30/45 Gallon Safety Cabinets
Catalog No. 19822445 Shop All Justrite Safety Group Products
Change view
Click to view available options
For Use With (Equipment):
30/45 gal. Cabinets
Catalog No. For Use With (Equipment)
19-822-445 30/45 gal. Cabinets
1 options
Catalog No. 19-822-445 Supplier Justrite™ Supplier No. 29943
  • $427.50
    $307.65 / Each of 1
    Save $119.85
    28% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Increase workplace safety and improve cleaniliness with the proper safety and storage equipment. The Justrite™ Spillslope Steel Dispensing Shelf for all 30/45 Gallon Safety Cabinets is a shelf with a shallow depth, which allows a dispensing can to be used on it.
  • Patented SpillSlope™ galvanized steel shelves direct spills to back and bottom of a leakproof sump.
  • Fits 30 & 45 gal. cabinets
Specifications
For Use With (Equipment) 30/45 gal. Cabinets
Material Steel
Width (English) Exterior 39.375 in.
Depth (English) Exterior 11 in.