Learn More
Justrite™ SpillSlope™ Steel Shelf for 22 Gallon Undercounter Safety Cabinet
Catalog No. 19822488 Shop All Justrite Safety Group Products
Change view
Click to view available options
For Use With (Equipment):
22 gal. Undercounter Cabinet
Catalog No. For Use With (Equipment)
19-822-488 22 gal. Undercounter Cabinet
1 options
Catalog No. 19-822-488 Supplier Justrite™ Supplier No. 29939
  • $220.00
    $149.65 / Each of 1
    Save $70.35
    32% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Increase the efficiency and space of existing cabinetry. The Justrite™ SpillSlope™ Steel Shelf for 22 Gallon Undercounter Safety Cabinet adds additional storage to cabinets.
  • Patented SpillSlope™ galvanized steel shelves direct spills to back and bottom of a leakproof sump. They adjust on 3 in. (76-mm) centers for versatile storage
  • Fits 22 gal. Undercounter cabinet
Specifications
For Use With (Equipment) 22 gal. Undercounter Cabinet
Depth (English) Exterior 18 in.
Material Steel
Width (English) Exterior 31.56 in.